Learn More
PHF11 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand:  Abnova H00051131-AP21.1Set
Product Code. 16143384
  • € 843.00 / 1 set
Specifications
PHF11
Unconjugated
APY/BCAP/IGEL/IGER/IGHER/NY-REN-34/NYREN34
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PHF11 (100 ug) 2. Detection antibody: mouse purified polyclonal anti-PHF11 (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
PHD finger protein 11
PHF11
RUO
51131
Human
Ab Pair