missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110827
This item is not returnable.
View return policy
Description
Immunogen sequence: TRMPRKTKNS YWQCSECDQA GSSDMEADMA METLPDGTKR SRRQIKEPVK FVPQDVPPEP KKIPIRNTRT RGRKRSFVPE EEKHEERVPR ERRQRQSVLQ KKPKAEDLR Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
PHF14 gene ontology annotations related to this gene include metal ion binding.
Specifications
| PHF14 | |
| Polyclonal | |
| Unconjugated | |
| PHF14 | |
| 1110001C23Rik; 4932409F11Rik; 5730446A07Rik; AA623952; AV297001; KIAA0783; mKIAA0783; PHD finger protein 14; Phf14; RGD1563764 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94880 | |
| PHF14 | |
| Recombinant Protein corresonding to Human PHF14. Recombinant protein control fragment (Product #RP-108313). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction