Learn More
PHF14 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. p-7210941 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16823285 20 μL 20µL
16813285 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16823285 Supplier Proteintech Supplier No. 247871AP20UL
Rabbit Polyclonal Antibody
PHF14 gene ontology annotations related to this gene include metal ion binding.
Specifications
Antigen PHF14
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PHF14
Gene Accession No. O94880
Gene Alias KIAA0783, PHD finger protein 14, PHF14
Gene Symbols PHF14
Host Species Rabbit
Immunogen PHF14 Fusion Protein Ag20542
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9678
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG