missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19796
This item is not returnable.
View return policy
Description
PHF16 Polyclonal specifically detects PHF16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PHF16 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| MGC138748, MGC138749, PHD finger protein 16KIAA0215JADE3, protein Jade-3 | |
| Rabbit | |
| 94 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.62mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q92613 | |
| PHF16 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PHF16. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9767 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction