missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF17 Monoclonal Antibody (658005)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561645

Product Code. 30534765

  • € 482.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Scaffold subunit of some HBO1 complexes, which have a histone H4 acetyltransferase activity (PubMed:16387653, PubMed:19187766, PubMed:20129055, PubMed:24065767). Plays a key role in HBO1 complex by directing KAT7/HBO1 specificity towards histone H4 acetylation (H4K5ac, H4K8ac and H4K12ac), regulating DNA replication initiation, regulating DNA replication initiation (PubMed:20129055, PubMed:24065767). May also promote acetylation of nucleosomal histone H4 by KAT5 (PubMed:15502158). Promotes apoptosis (PubMed:16046545). May act as a renal tumor suppressor (PubMed:16046545). Negatively regulates canonical Wnt signaling; at least in part, cooperates with NPHP4 in this function (PubMed:22654112). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PHF17
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q6IE81
JADE1
E. coli-derived recombinant human JADE-1/PHF-17, Met1-Ser137.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot, Immunocytochemistry
658005
Unconjugated
JADE1
AU041499; D530048A03Rik; gene for apoptosis and differentiation in epithelia; jade family PHD finger 1; jade family PHD finger protein 1; Jade1; Jade1 PHD finger protein; KIAA1807; mKIAA1807; PHD finger protein 17; PHD protein Jade-1; Phf17; Protein Jade-1
Mouse
Protein A/G
RUO
79960
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.