missing translation for 'onlineSavingsMsg'
Learn More

PHF2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB300-162APC

Product Code. 18514603

  • € 508.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PHF2 Polyclonal antibody specifically detects PHF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PHF2
Polyclonal
Western Blot
GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2
Internal peptide residues made to the 120 kDa PHF protein.
0.1 mL
Neuroscience
5253
Store at 4C in the dark.
IgG
Western Blot
APC
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.