missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98813B
This item is not returnable.
View return policy
Description
PHF2 Polyclonal antibody specifically detects PHF2 in Human samples. It is validated for Western Blot
Specifications
| PHF2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PHF2. | |
| 0.1 mL | |
| Neuroscience | |
| 5253 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction