missing translation for 'onlineSavingsMsg'
Learn More

PHF20L1 Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene

Product Code. 17502304
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17502304 100 μL 100µL
17522304 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17502304 Supplier Origene Technologies Supplier No. TA809760

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The exact function of PHF20L1 remains unknown.

Specifications

Antigen PHF20L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PHF20L1
Gene Accession No. A8MW92
Gene Alias CGI-72, TDRD20B, URLC1
Gene Symbols PHF20L1
Host Species Mouse
Immunogen Full length human recombit protein of human PHF20L1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51105
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.