missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF21A Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04043AF700
This item is not returnable.
View return policy
Description
PHF21A Polyclonal antibody specifically detects PHF21A in Human samples. It is validated for Western Blot
Specifications
| PHF21A | |
| Polyclonal | |
| Western Blot | |
| BHC80a, BHC80BM-006, BRAF35-HDAC complex protein BHC80, KIAA1696BRAF35/HDAC2 complex (80 kDa), PHD finger protein 21A | |
| The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_001095272.1 (GeneID 51317). | |
| 0.1 mL | |
| Neuroscience | |
| 51317 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction