missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHF21A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15712761 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15712761 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15712761 Supplier Invitrogen™ Supplier No. PA554829

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QEREQLEQKV KQLSNSISKC MEMKNTILAR QKEMHSSLEK VKQLIRLIHG IDLSKPVDSE ATVGAISNGP DCTPPANAAT STPAPSPSSQ SCTANCNQG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 93%.

PHF21A (PHD finger protein 21A) is a component of the BHC complex, a corepressor complex that represses transcription of neuron-specific genes in non-neuronal cells. The BHC complex is recruited at RE1/NRSE sites by REST and acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier. In the BHC complex, PHF21A may act as a scaffold.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF21A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Phf21a
Gene Accession No. Q96BD5
Gene Alias 80kDa; Bhc; Bhc80; BHC80a; BM-006; Braf35/HDAC complex (Bhc); BRAF35/HDAC2 complex; BRAF35/HDAC2 complex (80 kDa); BRAF35-HDAC complex protein BHC80; D030065N23Rik; KIAA1696; mBHC80; PFTF1; PHD finger protein 21A; Phf21a; RGD1560612
Gene Symbols Phf21a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF21A. Recombinant protein control fragment (Product #RP-93601).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.