missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHF21B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140271
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QKLLQRGSEL QNEHQQLEER DRRLASAVQK CLELKTSLLA RQRGTQSSLD R Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.
PHF21B (PHD finger protein 21B), also known as PHF4, is a 531 amino acid protein that contains one PHD-type zinc finger and is expressed as multiple alternatively spliced isoforms. The gene encoding PHF21B maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia.
Specifications
| PHF21B | |
| Polyclonal | |
| Unconjugated | |
| PHF21B | |
| A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EK2 | |
| PHF21B | |
| Recombinant protein corresponding to Human PHF21B. Recombinant protein control fragment (Product #RP-109423). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido