missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51062-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in the regulation of serine proteinases present in the brain or extravasated from the blood. Inhibitor of cathepsin G, kallikrein-8 and thrombin. May play an important role in the inner ear in the protection against leakage of lysosomal content during stress and loss of this protection results in cell death and sensorineural hearing loss.Specifications
| PHF6 | |
| Polyclonal | |
| Unconjugated | |
| PHF6 | |
| BORJ, KIAA1823, PHD finger protein 6, PHD like zinc finger protein, PHF6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 70998, 84295 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IWS0, Q9D4J7 | |
| Phf6 | |
| PHF6 Fusion Protein Ag2102 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction