Learn More
PHF7 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505156S
Encompass_Preferred
 View more versions of this product
Product Code. 17583262
  • € 242.00 / 30µL
Description
PHF7 Monoclonal antibody specifically detects PHF7 in Human samples. It is validated for Western Blot
Specifications
Specifications
PHF7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BWX1
Mouse
Affinity chromatography
RUO
51533
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1C2
Unconjugated
PHF7
1700006H01Rik; 1700010P14Rik; AI427892; AW555949; HSPC045; HSPC226; NYD-SP6; PHD finger protein 7; PHF7; testicular secretory protein Li 34; testis development protein NYD-SP6
Human recombinant protein fragment corresponding to amino acids 155-381 of human PHF7 produced in E.coli.
30 μL