missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14719-1-AP-20UL
This item is not returnable.
View return policy
Description
May regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane.Specifications
| PHGDH | |
| Polyclonal | |
| Unconjugated | |
| PHGDH | |
| 3 PGDH, 3PGDH, PDG, PGAD, PGD, PGDH, PGDH3, PHGDH, phosphoglycerate dehydrogenase, SERA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 236539, 26227, 58835 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08651, O43175, Q61753 | |
| PHGDH | |
| PHGDH Fusion Protein Ag6445 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction