missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHKA1/PHKA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106488
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KPB1/2.
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.
Specifications
| PHKA1/PHKA2 | |
| Polyclonal | |
| Unconjugated | |
| Phka2 | |
| 5330411D17; 6330505C01Rik; 9830108K24Rik; D330034O08; GSD9A; Pcyt1b; PHK; PHKA; Phka1; PHKA2; PhK-alpha-subunit; PHKLA; phosphorylase B kinase alpha subunit; phosphorylase b kinase regulatory subunit alpha liver isoform; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; phosphorylase kinase; phosphorylase kinase alpha 1; phosphorylase kinase alpha 2; phosphorylase kinase alpha L subunit; Phosphorylase kinase alpha M subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase regulatory subunit alpha 1; phosphorylase kinase regulatory subunit alpha 2; phosphorylase kinase, alpha 1; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis; phosphorylase kinase, alpha 2; phosphorylase kinase, alpha 2 (liver); PYK; PYKL; XLG; XLG2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110094, 18679, 5255, 5256, 64561, 678739 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18826, P46019, P46020, Q64649, Q8BWJ3 | |
| PHKA1, Phka2 | |
| A synthesized peptide derived from human PHKA2(Accession P46019), corresponding to amino acid residues L275-E325. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction