missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHKB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599686
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PHKB.
Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PHKD. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
Specifications
| PHKB | |
| Polyclonal | |
| Unconjugated | |
| PHKB | |
| AI463271; Phkb; Phosphorylase B; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase; phosphorylase kinase beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; phosphorylase kinase regulatory subunit beta; phosphorylase kinase subunit beta; phosphorylase kinase, beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 102093, 361377, 5257 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7TSH2, Q93100 | |
| PHKB | |
| A synthesized peptide derived from human PHKB(Accession Q93100), corresponding to amino acid residues V206-G256. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction