missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PHKG2 Antibody (OTI1F9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73373AF488
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PHKG2 Monoclonal specifically detects PHKG2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Tekniske data
| PHKG2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PHKG2 | |
| Recombinant protein fragment corresponding to amino acids 1-216 of human PHKG2 (NP_000285) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1F9 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.19, GSD9C, PHK-gamma-T, phosphorylase b kinase gamma catalytic chain, testis/liver isoform, Phosphorylase kinase subunit gamma-2, phosphorylase kinase, gamma 2 (testis), Phosphorylase kinase, gamma 2 (testis/liver), PSK-C3 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 5261.0 | |
| Store at 4°C in the dark. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion