missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKG2 Antibody (OTI1F9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73373UV
This item is not returnable.
View return policy
Description
PHKG2 Monoclonal specifically detects PHKG2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PHKG2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PHKG2 | |
| Recombinant protein fragment corresponding to amino acids 1-216 of human PHKG2 (NP_000285) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1F9 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.19, GSD9C, PHK-gamma-T, phosphorylase b kinase gamma catalytic chain, testis/liver isoform, Phosphorylase kinase subunit gamma-2, phosphorylase kinase, gamma 2 (testis), Phosphorylase kinase, gamma 2 (testis/liver), PSK-C3 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 5261.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction