Learn More
PHLDB2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-36546DL594
Product Code. 18710110
  • € 633.00 / 0.10mL
Description
PHLDB2 Polyclonal antibody specifically detects PHLDB2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PHLDB2
Polyclonal
50mM Sodium Borate
90102
Purified
Western Blot
DyLight 594
pleckstrin homology-like domain, family B, member 2
Store at 4°C in the dark.