missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHLDB2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17180944 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17180944 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180944 Supplier Invitrogen™ Supplier No. PA5113087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHLDB2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Seems to be involved in the assembly of the postsynaptic apparatus. May play a role in acetyl-choline receptor (AChR) aggregation in the postsynaptic membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLDB2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PHLDB2
Gene Accession No. Q86SQ0, Q8K1N2
Gene Alias AV253284; C820004H04Rik; FLJ21791; LL5 beta; LL5b; LL5beta; Phldb2; pleckstrin homology like domain family B member 2; pleckstrin homology-like domain family B member 2; pleckstrin homology-like domain, family B, member 2; protein LL5-beta
Gene Symbols PHLDB2
Host Species Rabbit
Immunogen Recombinant Human Pleckstrin homology-like domain family B member 2 protein (734-1030AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208177, 90102
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.