missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ phoP Polyclonal Antibody
Description
phoP Polyclonal Antibody for Western Blot, ELISA
Member of the two-component regulatory system PhoP/PhoQ involved in adaptation to low Mg(2+) environments and the control of acid resistance genes. In low periplasmic Mg(2+), PhoQ phosphorylates PhoP, resulting in the expression of PhoP-activated genes (PAG) and repression of PhoP-repressed genes (PRG). In high periplasmic Mg(2+), PhoQ dephosphorylates phospho-PhoP, resulting in the repression of PAG and may lead to expression of some PRG (By similarity). Mediates magnesium influx to the cytosol by activation of MgtA. Promotes expression of the two-component regulatory system rstA/rstB and transcription of the hemL, mgrB, nagA, slyB, vboR and yrbL genes.
Specifications
Specifications
| Antigen | phoP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | phoP |
| Gene Accession No. | P23836 |
| Gene Alias | phoP; Transcriptional regulatory protein PhoP |
| Gene Symbols | phoP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?