missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosducin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | Phosducin |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232661
|
Novus Biologicals
NBP2-56992 |
100 μL |
€ 624.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654178
|
Novus Biologicals
NBP2-56992-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosducin Polyclonal specifically detects Phosducin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Phosducin | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 5132 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DWRKFKLESQDSDSIPPSKKEILRQMSSPQSRNGKDSKERVSRKMSIQEYELI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| Human | |
| 33 kDa phototransducing protein, 33kDA phototransducing protein, G beta gamma binding protein, PHD, PhLP, phosducin, phosducin-like orphan protein | |
| PDC | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title