Learn More
Melford Phosphatase Inhibitor Cocktail II
Description
Lyophillized blend of five phosphatase inhibitors for use with animal tissue.
Lyophillized blend of five phosphatase inhibitors for use with animal tissue. A431 and Jurkat cell extracts. Suitable for extracts containing detergents. Reconstitute with 1ml water to produce a 100X concentrate. Dilute 1:100 before using.
Contains:
Imidazole
Sodium Fluoride
Sodium Molybdate
Sodium Orthovanadate
Sodium Tartrate, dihydrate
Specificity:
Protein Tyrosine Phoshatase
Acid Phosphatases
Alkaline Phosphatases
Specifications
Specifications
| Quantity | 1 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.