missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-14-3-3 sigma (Ser186) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564811
This item is not returnable.
View return policy
Description
Phospho-14-3-3 sigma (Ser186) Polyclonal Antibody detects endogenous levels of 14-3-3 sigma only when phosphorylated at Ser186.
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
Specifications
| Phospho-14-3-3 sigma (Ser186) | |
| Polyclonal | |
| Unconjugated | |
| SFN | |
| 14-3-3 protein sigma; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3sigma; epithelial cell marker protein 1; Er; HGNC:10773; HME1; Mkrn3; Mme1; SFN; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; Ywhas | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2810, 55948 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70456, P31947 | |
| SFN | |
| A synthesized peptide derived from human 14-3-3 sigma (Phospho-Ser186). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction