missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-14-3-3 zeta (Ser58) Polyclonal Antibody

Phospho-14-3-3 zeta (Ser58) Polyclonal Antibody
Brand: Invitrogen PA5104934
This item is not returnable.
View return policy
Description
Phospho-14-3-3 zeta (Ser58) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| Phospho-14-3-3 zeta (Ser58) | |
| Polyclonal | |
| Unconjugated | |
| Ywhaz | |
| 1110013I11Rik; 14-3-3 delta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 zeta; 1433Z; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 143Z; AI596267; AL022924; AU020854; epididymis luminal protein 4; epididymis secretory protein Li 3; epididymis secretory protein Li 93; factor activating exoenzyme S; FAS; HEL4; HEL-S-3; HEL-S-93; KCIP-1; mitochondrial import stimulation factor S1 subunit; Msfs1; phospholipase A2; protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; SEZ-2; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; tyrosine 3/tryptophan 5-monooxygenase activation protein zeta polypeptide; tyrosine 3-monooxygenase tryptophan 5-monooxygenase activation protein; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, delta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide; YWHAD; YWHAZ | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 22631, 25578, 7534 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P63101, P63102, P63104 | |
| Ywhaz | |
| A synthesized peptide derived from human YWHAZ(Accession P63104), corresponding to amino acid residues around phosphorylated Ser58. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction