Learn More
Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Recombinant Rabbit Monoclonal Antibody (4EB1T37T46-A5)

Rabbit Recombinant Monoclonal Antibody
€ 332.00 - € 562.00
Specifications
| Antigen | Phospho-4EBP1 (Thr37, Thr46) |
|---|---|
| Clone | 4EB1T37T46-A5 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15927204
|
Invitrogen™
MA527999 |
200 μL |
€ 562.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17116528
![]() |
Invitrogen™
MA536935 |
20 μL |
€ 332.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.Specifications
| Phospho-4EBP1 (Thr37, Thr46) | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13541, Q60876, Q62622 | |
| 116636, 13685, 1978 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr37/46 of human phospho 4E-BP1. | |
| Antibody |
| 4EB1T37T46-A5 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG κ | |
| Human, Mouse, Rat | |
| EIF4EBP1 | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| EIF4EBP1 | |
| Primary | |
| Protein A/G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.