missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Afadin (Ser1799) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104569
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Afadin only when phosphorylated at Ser1799.
AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.
Specifications
| Phospho-Afadin (Ser1799) | |
| Polyclonal | |
| Unconjugated | |
| Afdn | |
| 5033403D15Rik; Af6; Af-6; Afadin; Afadin adherens junction formation factor; afadin, adherens junction formation factor; afadin; afadin, adherens junction formation factor; AFDN; ALL1-fused gene from chromosome 6 protein; FLJ34371; Gm314; I-afadin; MLL-AF6; MLLT4; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; protein Af-6; RP3-431P23.3; RP3-470B24.4; S-afadin | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 4301 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P55196 | |
| Afdn | |
| A synthesized peptide derived from human AFDN(Accession P55196), corresponding to amino acid residues around phosphorylated Ser1799. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction