Learn More
Invitrogen™ Phospho-AGO2 (Tyr393) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143648
Description
This antibody was cross-absorbed to unphospho-Ago2 (Tyr-393), before affinity purification using phospho-Ago2 (Tyr-393) peptide. The antibody detects a 97 kDa* protein corresponding to the apparent molecular mass of phosphorylated Ago2 on SDS-PAGE immunoblots of human A431 cells that were treated with EGF. This reactivity is not observed after lambda phosphatase dephosphorylation.
Ago proteins are broadly expressed in somatic cells, associate with miRNAs and are key actors in different RNA silencing pathways. However, only the Ago2 protein displays endonucleolytic or 'slicer activity and can therefore execute miRNA-directed cleavage of target mRNA, provided that the base-pairing between the Ago2-associated miRNA and the mRNA sequence is perfect. In case of partial complementarity, the Ago2 protein fails to cleave, but instead interferes with translation of the target mRNA via its translational repression activity. Ago2 has been shown to play a non-redundant role in small RNA guided gene silencing processes, including RNA interference, translation repression and heterochromatinization. As a core element of the RISC complex, AGO2 initiates the degradation of target mRNAs through its catalytic activity in gene silencing processes guided by siRNAs or miRNAs. In addition to mRNA degradation or gene silencing guided by miRNAs, Ago2 also acts as a RNA slicer in a Dicer-independent way, as well as a regulator of miRNA maturation. Over-expression of Ago2 has been correlated with several aspects of cancers, including tumor cell growth and the overall survival of cancer patients. Furthermore, gene disruption in the mouse demonstrated that the Ago2 protein is essential for embryonic development and a key regulator of B-lymphoid and erythroid development.
Specifications
| Phospho-AGO2 (Tyr393) | |
| Polyclonal | |
| Unconjugated | |
| AGO2 | |
| 1110029L17Rik; 2310051F07Rik; AGO1; AGO2; AI225898; AL022874; argonaute 2; argonaute 2, RISC catalytic component; argonaute RISC catalytic component 2; argonaute RISC catalytic subunit 2; Argonaute1; argonaute2; argonaute2 {ECO:0000255; AW546247; CTA-204B4.6; eIF2C; eIF2C 1; eIF-2C 1; eIF2C 2; eIF-2C 2; eIF2C 2 {ECO:0000255; eIF-2C 2 {ECO:0000255; EIF2C1; Eif2c2; ENSMUSG00000072493; Eukaryotic translation initiation factor 2C 2; eukaryotic translation initiation factor 2C 2 {ECO:0000255; eukaryotic translation initiation factor 2C, 2; GERp95; Gm10365; Golgi ER protein 95 kDa; hAgo1; hAgo2; HAMAP-Rule:MF_03031}; Kiaa4215; mAgo2; mKIAA4215; PAZ Piwi domain protein; Piwi/Argonaute family protein meIF2C2; PPD; Protein argonaute-1; protein argonaute-2; Protein slicer; Q10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 239528, 27161, 59117 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q8CJG0, Q9QZ81, Q9UKV8 | |
| AGO2 | |
| Phospho-Ago2 (Tyr-393) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding tyrosine 393 in human Ago2. This peptide sequence is highly conserved in mouse, rat, chick, and fish Ago2. The site is also conserved in Ago1 and Ago4, but not in Ago3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.