missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-ALOX5 (Ser663) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15792806
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15792806 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15792806 Supplier Invitrogen™ Supplier No. PA564745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-ALOX5 (Ser663) Polyclonal Antibody detects endogenous levels of ALOX5 only when phosphorylated at Ser663.

This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ALOX5 (Ser663)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALOX5
Gene Accession No. P09917, P12527, P48999
Gene Alias 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; F730011J02; leukotriene A4 synthase; LOG5; LOW QUALITY PROTEIN: arachidonate 5-lipoxygenase; lox5; LOX-5; LOX5A; polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3
Gene Symbols ALOX5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALOX5 (Phospho-Ser663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11689, 240, 25290
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.