missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-ATF2 (Thr71) Recombinant Rabbit Monoclonal Antibody (ATF2T71-G3), FITC

Product Code. 15957194 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
Unit Size:
Pack of 10
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15957194 100 Tests Pack of 100
17136538 10 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15957194 Supplier Invitrogen™ Supplier No. MA527991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 interacts with a variety of viral oncoproteins and cellular tumor suppressors and is a target of the SAPK/JNK and p38 MAP kinase signaling pathways. Various forms of cellular stress, including genotoxic agents, inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Cellular stress activates ATF-2 by phosphorylation of Thr69 and Thr71. Both SAPK and p38 MAPK have been shown to phosphorylate ATF-2 at these sites in vitro and in cells transfected with ATF-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ATF2 (Thr71)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone ATF2T71-G3
Conjugate FITC
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Atf2
Gene Accession No. P15336, P16951
Gene Alias Activating 2; Activating transcription factor 2; activating transcription factor 2 splice variant ATF2-var2; ATF2; Atf-2; cAMP response element-binding protein CRE-BP1; cAMP responsive element binding protein 2, formerly; cAMP-dependent transcription factor ATF-2; CREB2; CREB-2; CRE-BP; CREBP1; CRE-BP1; Cyclic AMP-dependent transcription factor ATF-2; cyclic AMP-responsive element-binding protein 2; D130078H02Rik; D18875; HB16; Histone acetyltransferase ATF2; MGC111558; mXBP; MXBP protein; OTTHUMP00000205345; OTTHUMP00000205347; OTTHUMP00000205348; OTTHUMP00000205350; OTTHUMP00000205352; OTTHUMP00000205353; OTTHUMP00000205354; Tg(Gzma-Klra1)7Wum; TREB7
Gene Symbols Atf2
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Thr71 of human phospho ATF2.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11909, 1386
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.