Learn More
Invitrogen™ Phospho-ATG14 (Thr429) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5105669
Product Code. 16622115
  • € 500.00 / 100µL
Description
Antibody detects endogenous levels of Atg14 only when phosphorylated at Thr429.
ATG14 (Autophagy related protein homolog 14) is encoded by the ATG14 gene and is located on chromosome 14 in humans. It is also known as ATG14L or BARKOR (Beclin 1-associated autophagy-related key regulator). Autophagy is an evolutionarily conserved process that involves recycling of misfolded cellular proteins and degradation of dysfunctional organelles. Autophagy is induced as a cellular response to nutrient stress which includes the formation of autophagosomes, fusion of these with the lysosome and formation of the autophagolysosome. The Class III PI3-Kinase, Vps34 and its interacting partner Beclin 1 have been shown to form a complex which specifically includes ATG 14 (type 1) or UVRAG (type 2). Atg 14 guides the type 1 complex to the preautophagosomal structure (PAS) and is critical for the initiation of autophagosome formation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-ATG14 (Thr429)
Polyclonal
Unconjugated
Atg14
4832427M01; Atg14; ATG14 autophagy related 14 homolog; Atg14L; Atg14-like protein; autophagy related 14; autophagy-related protein 14-like protein; Bakor; Barkor; beclin 1-associated autophagy-related key regulator; Beclin 1-Interacting protein; D14Ertd114e; D14Ertd436e; Kiaa0831; RGD1304610; VATG14 autophagy related 14 homolog
Rabbit
Sequential chromatography
RUO
100504663, 22863, 305831
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
D4A4K3, Q6ZNE5, Q8CDJ3
Atg14
A synthesized peptide derived from human ATG14(Accession Q6ZNE5), corresponding to amino acid residues around phosphorylated Thr429.
100 μL
Primary
Human, Mouse, Rat