missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATRIP (Ser68) Polyclonal Antibody

Phospho-ATRIP (Ser68) Polyclonal Antibody
Brand: Invitrogen PA5105019
This item is not returnable.
View return policy
Description
Phospho-ATRIP (Ser68) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-ATRIP (Ser68) | |
| Polyclonal | |
| Unconjugated | |
| ATRIP | |
| 6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 235610, 301014, 84126 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BMG1, Q8WXE1 | |
| ATRIP | |
| A synthesized peptide derived from human ATRIP(Accession Q8WXE1), corresponding to amino acid residues around phosphorylated Ser68. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction