Learn More
Invitrogen™ Phospho-Bcl-3 (Ser41) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5105525
Product Code. 16601975
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of BCL-3 only when phosphorylated at Ser41.
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Bcl-3 (Ser41)
Polyclonal
Unconjugated
Bcl3
AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3
Rabbit
Sequential Chromatography
RUO
12051, 602
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P20749, Q9Z2F6
Bcl3
A synthesized peptide derived from human BCL3(Accession P20749), corresponding to amino acid residues around phosphorylated Ser41.