missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Beclin 1 (Ser90, Ser93, Ser96) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105647
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Beclin 1 only when phosphorylated at Ser90/93/96.
Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
Specifications
| Phospho-Beclin 1 (Ser90, Ser93, Ser96) | |
| Polyclonal | |
| Unconjugated | |
| BECN1 | |
| 4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 114558, 56208, 8678 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88597, Q14457, Q91XJ1 | |
| BECN1 | |
| A synthesized peptide derived from human BECN1(Accession Q14457), corresponding to amino acid residues around phosphorylated Ser90. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction