missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-beta Catenin/gamma Catenin (Tyr333, Tyr324) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17896948
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17896948 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17896948 Supplier Invitrogen™ Supplier No. PA5143665

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects a 92kDa* protein corresponding to the molecular mass of beta-Catenin on SDS-PAGE immunoblots of human A431 cells treated with pervanadate, but not in control cells. In addition, the pervanadate induced reactivity is not observed after alkaline phosphatase treatment.

beta-Catenin is a 92 kDa protein that binds to the cytoplasmic tail of E-Cadherin. The cadherins, transmembrane adhesion molecules, are found with catenins at adherens junctions. Deletions in the cytoplasmic domain of E-Cadherin eliminate catenin binding and result in a loss of cell adhesion. Tyrosine phosphorylation of beta-Catenin can regulate its interaction with critical components of adherens junctions. Both Fer and Fyn kinases phosphorylate tyrosine 142 in vitro. Overexpression of these kinases in epithelial cells disrupts interactions between alpha- and beta-Catenins. The phosphorylation of tyrosine 142 may act as a switch from the transcriptional to the adhesive role of beta-Catenin. Src family kinases can also phosphorylate tyrosine 86 and 654 in beta-Catenin. The Tyr-654 phosphorylation regulates beta-Catenin binding to E-cadherin. Thus, site-specific tyrosine phosphorylation of beta-Catenin may regulate protein-protein interactions leading to changes in cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-beta Catenin/gamma Catenin (Tyr333, Tyr324)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CTNNB1
Gene Accession No. P35222, Q02248, Q9WU82
Gene Alias Catenin beta1; CTNNB1
Gene Symbols CTNNB1
Host Species Rabbit
Immunogen Phospho-beta-Catenin (Tyr-333) synthetic peptide (coupled to KLH) that includes amino acid residues around tyrosine 333 of human beta-Catenin. This peptide sequence is highly conserved in rat and mouse beta-Catenin, and has high homology to the conserved site in gamma-Catenin (Tyr-324).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12387, 1499, 84353
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.