missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Beta Catenin (Ser552) Rabbit Polyclonal Antibody, Proteintech

Product Code. p-200107966 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235752 100 μL 100µL
30233366 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235752 Supplier Proteintech Supplier No. 287781AP100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Phospho-Beta Catenin (Ser552) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-Beta Catenin (Ser552). This antibody recognizes human, mouse, rat antigen. The Phospho-Beta Catenin (Ser552) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen Beta Catenin (Ser552)
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Polyclonal
Concentration 150 μg/mL
Conjugate Unconjugated
Gene Symbols CTNNB; DKFZp686D02253; FLJ25606; FLJ37923
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 86-100 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.