missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-beta Dystroglycan (Tyr892) Monoclonal Antibody (M117)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547624
This item is not returnable.
View return policy
Description
The antibody detects a 43 kDa* protein on SDS-PAGE immunoblots of human HepG2 or HeLa cells treated with pervanadate, but not in control cells.
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
Specifications
| Phospho-beta Dystroglycan (Tyr892) | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q14118, Q62165 | |
| DAG1 | |
| Clone (M117) was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 892 of human dystroglycan. This peptide sequence has high homology to the conserved tyrosine site in rat and mouse dystroglycan. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| M117 | |
| Unconjugated | |
| DAG1 | |
| 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 | |
| Mouse | |
| Protein A | |
| RUO | |
| 114489, 13138, 1605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction