missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Bim (Ser69) Monoclonal Antibody (K.724.4)
Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514814
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
BIM belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains a Bcl-2 homology domain 3 (BH3). It has been shown to interact with other members of the BCL-2 protein family, including BCL2, BCL2L1/BCL-X(L), and MCL1, and to act as an apoptotic activator. The expression of this gene can be induced by nerve growth factor (NGF), as well as by the forkhead transcription factor FKHR-L1, which suggests a role of this gene in neuronal and lymphocyte apoptosis. Transgenic studies of the mouse counterpart suggested that this gene functions as an essential initiator of apoptosis in thymocyte-negative selection. Several alternatively spliced transcript variants of this gene have been identified.
Specifications
| Phospho-Bim (Ser69) | |
| Monoclonal | |
| 185 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| O43521, O54918 | |
| BCL2L11 | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer69 of human Bim protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| K.724.4 | |
| Unconjugated | |
| BCL2L11 | |
| 1500006F24Rik; BAM; Bcl2 interacting mediator of cell death; bcl-2 interacting mediator of cell death; bcl-2 interacting protein Bim; BCL2 like 11; Bcl-2 like 11; Bcl-2 related apoptotic gene product BimL; bcl2-interacting mediator of cell death; BCL2L11; Bcl2-L-11; BCL2-like 11 (apoptosis facilitator); bcl-2-like protein 11; bcl-2-like protein 11-like; bcl-2-related ovarian death agonist; bcl-2-related ovarian death protein; BIM; Bim EL; Bim-A; BIM-alpha6; BIM-beta6; BIM-beta7; BIMEL; BimEL protein; BimL; BOD; OTTHUMP00000197013; OTTHUMP00000204247; Unknown (protein for MGC:134444) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10018, 12125 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction