missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Rabbit Monoclonal Antibody (23GB4495), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Phospho-C-Jun/D-Jun (Ser73, Ser100) |
|---|---|
| Clone | 23GB4495 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088797
![]() |
Invitrogen™
MA552704 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088519
|
Invitrogen™
MA552705 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Monoclonal antibody specifically detects Phospho-C-Jun/D-Jun (Ser73, Ser100) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Phospho-C-Jun/D-Jun (Ser73, Ser100) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P05412, P05627, P15066, P17325, P17535, P52909 | |
| 16476, 16478, 24516, 24518, 3725, 3727 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4495 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Activator Protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; CJUN; C-JUN; c-Jun protein; enhancer-binding protein AP1; fj36h07; immediate early; JUN; Jun A; Jun activation domain binding protein; jun D proto-oncogene; jun oncogene; jun proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; p39; Proto-Oncogene CJun; proto-oncogene c-Jun; Rjg-9; si:dkey-251j8.3; transcription factor; transcription factor AP-1; Transcription Factor AP-1 Subunit Jun; Transcription Factor Jun; Transcription factor jun-D; Unknown (protein for MGC:65863); V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog; wu:fj36h07; zgc:65863 | |
| A synthesized peptide derived from human Phospho-c-Jun (Ser73)+JunD (Ser100) (66-80AA). | |
| Primary | |
| -20°C | |
| JUN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title