missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Rabbit Monoclonal Antibody (K01_2K98)

Description
Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Monoclonal Antibody for Western Blot
C-Jun/D-Jun are members of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various cell stimuli, and targets specific transcription factors, and thus mediates immediate-early gene expression in response to cell stimuli. The activation of this kinase by tumor-necrosis factor alpha (TNF-alpha) is found to be required for TNF-alpha induced apoptosis. This kinase is also involved in UV radiation induced apoptosis, which is thought to be related to cytochrom c-mediated cell death pathway. Studies of the mouse counterpart of this gene suggested that this kinase play a key role in T cell proliferation, apoptosis and differentiation. Several alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq, Apr 2016].
Specifications
Specifications
| Antigen | Phospho-C-Jun/D-Jun (Ser73, Ser100) |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2K98 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | JUN |
| Gene Accession No. | P05412, P05627, P15066, P17325, P17535, P52909 |
| Gene Alias | Activator Protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; CJUN; C-JUN; c-Jun protein; enhancer-binding protein AP1; fj36h07; immediate early; JUN; Jun A; Jun activation domain binding protein; jun D proto-oncogene; jun oncogene; jun proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; p39; Proto-Oncogene CJun; proto-oncogene c-Jun; Rjg-9; si:dkey-251j8.3; transcription factor; transcription factor AP-1; Transcription Factor AP-1 Subunit Jun; Transcription Factor Jun; Transcription factor jun-D; Unknown (protein for MGC:65863); V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog; wu:fj36h07; zgc:65863 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?