missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Rabbit Monoclonal Antibody (K01_2K98)
GREENER_CHOICE

Product Code. p-200114430
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825603 100 μL
30825828 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825603 Supplier Invitrogen™ Supplier No. MA563977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-C-Jun/D-Jun (Ser73, Ser100) Recombinant Monoclonal Antibody for Western Blot

C-Jun/D-Jun are members of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various cell stimuli, and targets specific transcription factors, and thus mediates immediate-early gene expression in response to cell stimuli. The activation of this kinase by tumor-necrosis factor alpha (TNF-alpha) is found to be required for TNF-alpha induced apoptosis. This kinase is also involved in UV radiation induced apoptosis, which is thought to be related to cytochrom c-mediated cell death pathway. Studies of the mouse counterpart of this gene suggested that this kinase play a key role in T cell proliferation, apoptosis and differentiation. Several alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq, Apr 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-C-Jun/D-Jun (Ser73, Ser100)
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2K98
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene JUN
Gene Accession No. P05412, P05627, P15066, P17325, P17535, P52909
Gene Alias Activator Protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; CJUN; C-JUN; c-Jun protein; enhancer-binding protein AP1; fj36h07; immediate early; JUN; Jun A; Jun activation domain binding protein; jun D proto-oncogene; jun oncogene; jun proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; p39; Proto-Oncogene CJun; proto-oncogene c-Jun; Rjg-9; si:dkey-251j8.3; transcription factor; transcription factor AP-1; Transcription Factor AP-1 Subunit Jun; Transcription Factor Jun; Transcription factor jun-D; Unknown (protein for MGC:65863); V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog; wu:fj36h07; zgc:65863
Gene Symbols JUN, Jund
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser73 of human Jun.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16476, 16478, 24516, 24518, 3725, 3727
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.