Learn More
Phospho-c-MYC (Ser62) Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18009825 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18009825 20 μL 20µL
18027385 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18009825 Supplier Proteintech Supplier No. 289151AP20UL
Rabbit Polyclonal Antibody
The Phospho-c-MYC (Ser62) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-c-MYC (Ser62). This antibody recognizes human antigen. The Phospho-c-MYC (Ser62) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen Phospho-c-MYC (Ser62)
Applications Western Blot, ELISA
Concentration 650 μg/mL
Gene Symbols bHLHe39, c Myc, MRTL, MYC, Myc proto oncogene protein, Phopho-Ser62-MYC, Proto oncogene c Myc, Transcription factor p64
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 4609
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG