missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Calpain 2 (Ser50) Polyclonal Antibody

Phospho-Calpain 2 (Ser50) Polyclonal Antibody
Brand: Invitrogen PA5105976
This item is not returnable.
View return policy
Description
Phospho-Calpain 2 (Ser50) Polyclonal Antibody for Western Blot
Specifications
| Phospho-Calpain 2 (Ser50) | |
| Polyclonal | |
| Unconjugated | |
| CAPN2 | |
| 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 12334, 29154, 824 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08529, P17655, Q07009 | |
| CAPN2 | |
| A synthesized peptide derived from human CAPN2(Accession P17655), corresponding to amino acid residues around phosphorylated Ser50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction