missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CaMKI (Thr177) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105644
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of CaMK1 alpha only when phosphorylated at Thr177.
CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Specifications
| Phospho-CaMKI (Thr177) | |
| Polyclonal | |
| Unconjugated | |
| CAMK1 | |
| AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 171503, 52163, 8536 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14012, Q63450, Q91YS8 | |
| CAMK1 | |
| A synthesized peptide derived from human CAMK1(Accession Q14012), corresponding to amino acid residues around phosphorylated Thr177. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction