missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Cdc20 (Thr70) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105704
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of CDC20 only when phosphorylated at Thr70.
Required for full ubiquitin ligase activity of the anaphase promoting complex/cyclosome (APC/C) and may confer substrate specificity upon the complex. Is regulated by MAD2L1: in metaphase the MAD2L1-CDC20-APC/C ternary complex is inactive and in anaphase the CDC20-APC/C binary complex is active in degrading substrates. The CDC20-APC/C complex positively regulates the formation of synaptic vesicle clustering at active zone to the presynaptic membrane in postmitotic neurons. CDC20-APC/C-induced degradation of NEUROD2 induces presynaptic differentiation.
Specifications
| Phospho-Cdc20 (Thr70) | |
| Polyclonal | |
| Unconjugated | |
| CDC20 | |
| 2310042N09Rik; bA276H19.3; C87100; cdc20; CDC20 cell division cycle 20 homolog; cdc20.S; CDC20A; cell cycle protein p55CDC; cell division cycle 20; cell division cycle 20 homolog; cell division cycle 20 S homeolog; cell division cycle protein 20 homolog; fizzy1; mmCdc20; p55cdc; XELAEV_18025163mg; X-FZY | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 107995, 64515, 991 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12834, Q62623, Q9JJ66 | |
| CDC20 | |
| A synthesized peptide derived from human CDC20(Accession Q12834), corresponding to amino acid residues around phosphorylated Thr70. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction