Learn More
Invitrogen™ Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (Chk1S317-F10)

Rabbit Recombinant Monoclonal Antibody
€ 331.00 - € 562.00
Specifications
| Antigen | Phospho-Chk1 (Ser317) |
|---|---|
| Clone | Chk1S317-F10 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15997184
|
Invitrogen™
MA527985 |
200 μL |
€ 562.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17176588
![]() |
Invitrogen™
MA537001 |
20 μL |
€ 331.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.Specifications
| Phospho-Chk1 (Ser317) | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14757, O35280 | |
| 1111, 12649 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1. | |
| Antibody |
| Chk1S317-F10 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG κ | |
| Human, Mouse | |
| Chek1 | |
| C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W | |
| Chek1 | |
| Primary | |
| Protein A/G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.