missing translation for 'onlineSavingsMsg'
Learn More

Phospho-DAPK2 (Ser318) Polyclonal Antibody, Invitrogen™

Product Code. 15525391
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525391 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15525391 Supplier Invitrogen Supplier No. PA538466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-DAPK2 (Ser318)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DAPK2
Gene Accession No. Q9UIK4, Q8VDF3
Gene Alias DRP-1, DRP1, DAP kinase 2, DAP-kinase-related protein 1 beta isoform
Gene Symbols DAPK2
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human DAPK2 around the phosphorylation site of Ser318 (K-L-SP-F-S)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13143, 23604
Target Species Human, Mouse, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.