missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-DISC1 (Ser710) Polyclonal Antibody

Phospho-DISC1 (Ser710) Polyclonal Antibody
Brand: Invitrogen PA5143680
This item is not returnable.
View return policy
Description
Phospho-DISC1 (Ser710) Polyclonal Antibody for Western Blot
Specifications
| Phospho-DISC1 (Ser710) | |
| Polyclonal | |
| Unconjugated | |
| DISC1 | |
| C1orf136; DISC1; disrupted in schizophrenia 1; disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein; KIAA0457; RP4-730B13.1; SCZD9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 244667 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q811T9 | |
| DISC1 | |
| Phospho-DISC1 (Ser-710) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-710 in mouse DISC1. This peptide sequence is well conserved in rat DISC1 (Ser-703), but has five amino acid differences with the conserved site in human DISC1 (Ser-713). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction