missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-DNM1L (Ser637) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101038
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of DRP1 only when phosphorylated at Ser637.
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
Specifications
| Phospho-DNM1L (Ser637) | |
| Polyclonal | |
| Unconjugated | |
| DNM1L | |
| 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 10059, 114114, 74006 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00429, O35303, Q8K1M6 | |
| DNM1L | |
| A synthesized peptide derived from human DNM1L(Accession O00429), corresponding to amino acid residues around phosphorylated Ser637. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction