missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-CRMP1 (Tyr504) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105983
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of CRMP-1 only when phosphorylated at Tyr504.
Collapsin-response mediator proteins (CRMPs) are highly expressed in the developing brain where they play major roles in axonal outgrowth, neurite differentiation, and apoptosis. Their continued expression in areas of high synaptic remodeling such as the cerebellum, hippocampus, and the olfactory system suggests that these proteins may also be involved in adult brain plasticity. CRMP-1 was initially identified as a dihydro-pyrimidinase expressed exclusively in brain; later studies have shown that it is involved with neurotrophin (NT) 3-induced neurite formation and outgrowth. CRMP-1 localization switches from axonal to somatodendritic when neurons reach functional maturity, suggesting that it is involved in early neuronal differentiation as well as in later processes related to the survival or death of the newly generated neurons.
Specifications
| Phospho-CRMP1 (Tyr504) | |
| Polyclonal | |
| Unconjugated | |
| CRMP1 | |
| Collapsin response mediator protein 1; Crmp1; CRMP-1; dihydropyrimidinase-like 1; dihydropyrimidinase-related protein 1; Dpysl1; DRP1; DRP-1; Ulip3; ULIP-3; Unc-33-like phosphoprotein 3 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 12933, 1400, 25415 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97427, Q14194, Q62950 | |
| CRMP1 | |
| A synthesized peptide derived from human CRMP1(Accession Q14194), corresponding to amino acid residues around phosphorylated Tyr504. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction